Order Entry
Spain
ContactUsLinkComponent
Triphenylphosphine for synthesis, Sigma-Aldrich®
Health
Irritant
Triphenylphosphine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Health
Irritant
Triphenylphosphine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
     - 0.0001 g/l (25 °C)
  • Melting Point:
    78.5 - 81.5 °C
  • Molar Mass:
    262.29 g/mol
  • Bulk Density:
    500 - 600 kg/m3
  • Boiling Point:
    195 - 205 °C (7 hPa)
  • Vapor Pressure:
    0.01 hPa (88 °C)
  • Flash Point:
    182 °C
  • Density:
    1.194 g/cm3 (20 °C)
  • Ignition Point:
    425 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together