Triphenylphosphine
1
Catalog # CAYM705018-1
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
- CAS Number:603-35-0
- Formula:C₁₈H₁₅P
- MDL Number:MFCD00003043
- Molecular Weight:262.29 g/mol
- Merck Index:12,09873
- Boiling Pt:377 °C (1013 hPa)
- Melting Pt:79 °C
- Density:1.194 g/cm³ (20 °C)
- Flash Pt:180 °C
- Storage Temperature:Ambient
- EINECS:210-036-0
Specifications
Specifications
- Pk:1



