
Tetrabutylammonium iodide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:311-28-4
:
Formula:C₁₆H₃₆IN
MDL Number:MFCD00011636
:369.37 g/mol
Melting Pt:142…148 °C
Density:1.0027 g/cm³ (25 °C)
EINECS:206-220-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together