

Tributyl phosphate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:126-73-8
:
Formula:(CH₃(CH₂)₃O)₃PO
MDL Number:MFCD00009436
:266.32 g/mol
Merck Index:13,09692
Boiling Pt:289 °C (760 mmHg)
Melting Pt:–79 °C
Density:0.979 g/cm³
Flash Pt:146 °C
Storage Temperature:Ambient
EINECS:204-800-2
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4230 to 1.4250 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥99.0 %
- Color scale:≤50 APHA
Specification Test Results
Frequently Bought Together