Order Entry
Spain
ContactUsLinkComponent
(3-Mercaptopropyl)trimethoxysilane for synthesis, Sigma-Aldrich®
 
 
(3-Mercaptopropyl)trimethoxysilane for synthesis, Sigma-Aldrich®
  8.18818.0100
 :  Merck
 :  
 
 
(3-Mercaptopropyl)trimethoxysilane for synthesis, Sigma-Aldrich®
  8.18818.0100
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

 

 

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    196.34 g/mol
  • Boiling Point:
    213 - 215 °C (1013 hPa)
  • Vapor Pressure:
    0.2 hPa (20 °C)
  • Flash Point:
    85 °C
  • Refractive Index:
    1.4402 (25 °C, 589 nm)
  • Density:
    1.05 g/cm3 (25 °C)
  • Ignition Point:
    235 °C

 

 

 :
 :196.34 g/mol
 :215 °C (1013 hPa)
 :–50 °C
 :1.056 g/cm³ (20 °C)
 :>65 °C
 :Ambient
 :224-588-5
 :3082
 :9,III
 :M-Clarity™ quality level = MQ200
 :GHS07
 :GHS09
 :4420-74-0
 :HSH₂CCH₂CH₂Si(OCH₃)₃
 :MFCD00004901