Order Entry
Spain
ContactUsLinkComponent
Strontium carbonate
Strontium carbonate
Catalog # PROONIST-987
CAS Number:  
Strontium carbonate
Catalog # PROONIST-987
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Merck Index:14,08838
Boiling Pt:2647 °C (1 mmHg)
Melting Pt:1768.85 °C
Density:3.548 g/cm³ (20 °C)
 :147.63 g/mol
Storage Temperature:Ambient
EINECS:216-643-7
CAS Number:1633-05-2
Formula:CO₃Sr
MDL Number:MFCD00011250