Order Entry
Spain
ContactUsLinkComponent
Calcium carbonate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E170 (max. 0.0001% Al), precipitated, SAFC®
Calcium carbonate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E170 (max. 0.0001% Al), precipitated, SAFC®
Catalog # 1.12120.9029
CAS Number:  
Calcium carbonate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E170 (max. 0.0001% Al), precipitated, SAFC®
Catalog # 1.12120.9029
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C
Density:2.71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-439-9
CAS Number:471-34-1
Formula:CaCO₃
MDL Number:MFCD00010906