Order Entry
Spain
ContactUsLinkComponent
 
Calcium carbonate 98.5-100.5% (by anhydrous basis), Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP ,E170, precipitated, Fluka™
 
Calcium carbonate 98.5-100.5% (by anhydrous basis), Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP ,E170, precipitated, Fluka™

 

  • Assay (calculated to the dried substance):
    98.5 - 100.5 %
  • Insoluble in acetic acid:
    Max. 0.2 %
  • Insoluble in hydrochloric acid:
    Max. 0.2 %
  • Loss on drying (200°C):
    Max. 2.0 %
  • Magnesium and alkali metals:
    Max. 1.0 %
  • Arsenic (As):
    Max. 2 ppm
  • Barium (Ba):
    complying
  • Cadmium (Cd):
    Max. 1 ppm
  • Chromium (Cr):
    Max. 10 ppm
  • Copper (Cu):
    Max. 10 ppm
  • Iron (Fe):
    Max. 200 ppm
  • Mercury (Hg):
    Max. 0.5 ppm
  • Lead (Pb):
    Max. 3 ppm
  • Antimony (Sb):
    Max. 10 ppm
  • Zinc (Zn):
    Max. 10 ppm
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Sb, Cu, Cr, Zn, Ba:
    Max. 100 ppm
  • Chloride (Cl):
    Max. 200 ppm
  • Fluoride (F):
    Max. 50 ppm
  • Sulfate (SO₄):
    Max. 0.2 %
  • Residual solvents:
    complying

 

 

Frequently Bought Together