Sodium chlorite tech. ∼80%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-19-2
:
Formula:ClNaO₂
MDL Number:MFCD00003478
:90.44 g/mol
Merck Index:14,8600
Boiling Pt:112 °C (1013 hPa)
Melting Pt:180…200 °C (dec.)
Density:1.28 g/cm³ (20 °C)
EINECS:231-836-6
UN:1496
ADR:5.1,II
:GHS03
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together