

4,4'-Bis(diethylamino)benzophenone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:90-93-7
:
Formula:C₂₁H₂₈N₂O
MDL Number:MFCD00009044
:324.47 g/mol
Boiling Pt:420 °C (1013 hPa)
Melting Pt:96.5 °C
EINECS:202-025-4
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:90 - 95 °C
- Molar Mass:324.46 g/mol
- Bulk Density:400 kg/m3
- Flash Point:151 °C
- pH:4 - 6 (50 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together