


Thymol
Catalog # EDQMT1350000
Supplier: EDQM
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:150.22 g/mol
Merck Index:13,09476
Boiling Pt:233 °C (1013 hPa)
Melting Pt:49…51 °C
Density:0.965 g/cm³ (25 °C)
Flash Pt:104 °C
EINECS:201-944-8
UN:2430
ADR:8,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:89-83-8
Formula:2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
MDL Number:MFCD00002309