2,4-Dimethylphenylboronic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:55499-44-0
:
Formula:C₈H₁₁BO₂
MDL Number:MFCD02683101
Molecular Weight:149.98 g/mol
Melting Pt:204…208 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together