Order Entry
Spain
Orders LinkContactUsLinkComponent
Thymol blue
Thymol blue
Certificates
Thymol blue

2 Options Available Below

Product Details & Documents

CAS No.: 76-61-9 Molecular Formula: C27H30O5S Formula Weight: 466.60 Melting Point: 223° dec. Merck Reference: 14,9400 MDL No.: MFCD00005869

Synonyms:  Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-61-9
  • Formula:C₂₇H₃₀O₅S
  • MDL Number:MFCD00005869
  • Molecular Weight:466.6 g/mol
  • Merck Index:14,9400
  • Melting Pt:223 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-973-3
Product Family Options

Product Information