Order Entry
Spain
ContactUsLinkComponent
p-Toluenesulphonyl chloride for synthesis, Sigma-Aldrich®
Corrosive
Irritant
p-Toluenesulphonyl chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
p-Toluenesulphonyl chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) Hydrolysis
  • Melting Point:
    67 °C
  • Molar Mass:
    190.65 g/mol
  • Bulk Density:
    800 kg/m3
  • Boiling Point:
    135 °C (13 hPa)
  • Flash Point:
    128 °C
  • Density:
    1.49 g/cm3 (20 °C)
  • pH:
    (H2O) strongly acid
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

Specification Test Results

Related Information

Frequently Bought Together