Order Entry
Spain
ContactUsLinkComponent
3-Nitrotoluene
3-Nitrotoluene
Catalog # EHERC15615300
Supplier:  EHRENSTORFER
CAS Number:  
3-Nitrotoluene
Catalog # EHERC15615300
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
Merck Index:12,06748
Boiling Pt:230 °C (1013 hPa)
Melting Pt:15.5 °C
Density:1.1571 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-728-6
UN:1664
ADR:6.1,II
 :137.14 g/mol
CAS Number:99-08-1
Formula:C₇H₇NO₂
MDL Number:MFCD00007265