Order Entry
Spain
ContactUsLinkComponent
 
 
 
 
α,α,α-Trifluoro-p-cresol 98%
 
undefined
 
 
 
α,α,α-Trifluoro-p-cresol 98%

 

  • Appearance:
    White to yellow-brown Crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    40 to 46 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together