4-Acetamidobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:556-08-1
:
Formula:C₉H₉NO₃
MDL Number:MFCD00002534
:179.18 g/mol
Boiling Pt:399 °C (1013 hPa)
Melting Pt:∼ 261 °C (dec.)
Storage Temperature:Ambient
EINECS:209-114-7
:GHS07
Related Information
Frequently Bought Together