
Benzaldehyde
Catalog # PRLSMC307589.1000
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:106.12 g/mol
Merck Index:13,01057
Boiling Pt:179 °C (1013 hPa)
Melting Pt:–26 °C
Density:1.049 g/cm³ (20 °C)
Flash Pt:64 °C
Storage Temperature:Ambient
EINECS:202-860-4
UN:1990
ADR:9,III
:GHS07
CAS Number:100-52-7
Formula:C₆H₅CHO
MDL Number:MFCD00003299