Order Entry
Spain
ContactUsLinkComponent
2,2,6,6-Tetramethylpiperidine for synthesis, Sigma-Aldrich®
2,2,6,6-Tetramethylpiperidine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
2,2,6,6-Tetramethylpiperidine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    25.9 g/l (20 °C)
  • Melting Point:
    -59 °C
  • Molar Mass:
    141.25 g/mol
  • Boiling Point:
    152 - 156 °C
  • Vapor Pressure:
    14 hPa (39 °C)
  • Flash Point:
    37 °C
  • Density:
    0.83 g/cm3 (20 °C)
  • Ignition Point:
    290 °C

Specification Test Results

Related Information

Frequently Bought Together