Sunset yellow FCF 90% (dye content), Sigma-Aldrich®
465224-25G
:
- Pack type:Poly bottle
- Pk:25 g
- Seq:3308
- Infrared Spectrum:Conforms to Structure
- Wavelength in H₂O:479 - 485 nm
- Extinction Coefficient :≥20000
- Carbon:≥37.3%
- Nitrogen:≥5.4%
- Purity (HPLC):≥88.0%
:
:452.36 g/mol
:>300 °C (lit.)
:0,617 g/cm³ (26 °C)
:M-Clarity™ quality level = MQ200
:Room Temperature
:220-491-7
:2783-94-0
:C₁₆H₁₀N₂Na₂O₇S₂
:MFCD00036437