Order Entry
Spain
ContactUsLinkComponent
4-Methyl-2-pentyne ≥96.0% (by GC)
4-Methyl-2-pentyne ≥96.0% (by GC)
Catalog # TCIAM0270-5ML
Supplier:  TCI
CAS Number:  
undefined
4-Methyl-2-pentyne ≥96.0% (by GC)
Catalog # TCIAM0270-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :82.15 g/mol
Boiling Pt:72°C
Flash Pt:-4°C
Flash Pt:–4°C
UN:3295
ADR:3,II
 :GHS02
 :GHS08
CAS Number:21020-27-9
Formula:C₆H₁₀
MDL Number:MFCD00041615