Ethyl oleate
Supplier: Cayman Chemical
Jump to:
About this item
Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
- CAS Number:111-62-6
- Formula:C₂₀H₃₈O₂
- MDL Number:MFCD00009579
- Molecular Weight:310.52 g/mol
- Merck Index:12,06965
- Boiling Pt:386 °C (1013 hPa)
- Melting Pt:–32 °C
- Density:0.87 g/cm³ (20 °C)
- Flash Pt:174 °C
- Storage Temperature:Ambient
- EINECS:203-889-5
2 Options Available Below
Product Details & Documents
Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
- CAS Number:111-62-6
- Formula:C₂₀H₃₈O₂
- MDL Number:MFCD00009579
- Molecular Weight:310.52 g/mol
- Merck Index:12,06965
- Boiling Pt:386 °C (1013 hPa)
- Melting Pt:–32 °C
- Density:0.87 g/cm³ (20 °C)
- Flash Pt:174 °C
- Storage Temperature:Ambient
- EINECS:203-889-5



