Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methyl-2-butanol
2-Methyl-2-butanol ≥98%
2-Methyl-2-butanol ≥98%
:
Thermo Fisher Scientific
2-Methyl-2-butanol ≥98%
:
Thermo Fisher Scientific
:
75-85-4
:
:
C₅H₁₂O
:
MFCD00004478
:
88.15 g/mol
:
14,7140
:
101…103 °C
:
∼–12 °C
:
0.805
:
20 °C (68 °F)
:
Ambient
:
200-908-9
:
1105
:
3,II
:
GHS02
:
GHS07
msds_ALFAA18304
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99%, TECHNICAL
: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Dichloromethane, EMPARTA® ACS analytical reagent, Supelco®
: Merck
Methyl ethyl ketone ≥99%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Acetic anhydride for synthesis, Sigma-Aldrich®
: Merck
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Toluene-[D₈] 100.0 atom % D for NMR spectroscopy
: Thermo Fisher Scientific
Methyl acetate, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific
Ethyl (R)-(-)-3-hydroxybutyrate 98%
: Thermo Fisher Scientific