


Bis(2-ethylhexyl)amine for synthesis, Sigma-Aldrich®
Catalog # 8.18191.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 ml
- Seq:2590
Specifications
Specification Test Results
- Solubility:0.045 g/l (20 °C)
- Molar Mass:241.45 g/mol
- Boiling Point:122 - 124 °C (7 hPa)
- Vapor Pressure:0.01 hPa (20 °C)
- Flash Point:121 °C
- Explosion Limit:0.6 - 3.7 %(V)
- Density:0.80 g/cm3 (20 °C)
- pH:7 (H2O)
- Ignition Point:230 °C
Specification Test Results
About this item
:
Boiling Pt:281.1 °C (1013 hPa)
Melting Pt:< –60 °C
Density:0.8062 g/cm³ (20 °C)
EINECS:203-372-4
ADR:8 (6.1),II
:M-Clarity™ quality level = MQ200
Flash Pt:121 °C
:241.46 g/mol
UN:2922
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
CAS Number:106-20-7
Formula:C₁₆H₃₅N
MDL Number:MFCD00009489