L(-)-Glutathione (reduced form) ≥98%, high purity
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:70-18-8
:
Formula:C₁₀H₁₇N₃O₆S
MDL Number:MFCD00065939
:307.33 g/mol
Merck Index:13,04488
Melting Pt:192…195 °C
Density:1.475 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-725-4
Specification Test Results
- Iron:≤0.002%
- Lead:≤0.001%
- Loss on drying:≤1%
- Purity:≥98%
- Specific rotation:−18.5 to −15.5°
Specification Test Results
Frequently Bought Together