

Chloromethyl pivalate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18997-19-8
:
Formula:C₆H₁₁ClO₂
MDL Number:MFCD00000884
:150.6 g/mol
Boiling Pt:146…148 °C
Density:1.045
Flash Pt:40 °C (104 °F)
Storage Temperature:Ambient
EINECS:242-735-1
ADR:3,III
:GHS05
:GHS02
Related Information
Frequently Bought Together