


Sodium methoxide, anhydrous, powder, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:124-41-4
:
Formula:CH₃NaO
MDL Number:MFCD00012179
:54.02 g/mol
Melting Pt:126 °C (dec.)
Density:1,3 g/cm³ (20 °C)
EINECS:204-699-5
UN:1431
ADR:4.2,II
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow Powder
- Alkalinity:≥99 % (Total)
- Titration:≥97 %
- NaOH + Na2CO3 (as NaOH):≤2 %
- Sodium (Na):4.5 to 5.5 %
Specification Test Results
Frequently Bought Together