Order Entry
Spain
ContactUsLinkComponent
2-Ethoxyethanol ≥99%, TECHNICAL
2-Ethoxyethanol ≥99%, TECHNICAL
Catalog # 24043.294
Supplier:  VWR Chemicals
CAS Number:  
undefined
2-Ethoxyethanol ≥99%, TECHNICAL
Catalog # 24043.294
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Refractive index (20°C):
    1.405 - 1.408

Specification Test Results

About this item

 :
 :90.12 g/mol
Merck Index:13,03786
Boiling Pt:135 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.932 g/cm³ (20 °C)
Flash Pt:40 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-804-1
UN:1171
ADR:3,III
REACH:01-2119560582-38
 :GHS02
 :GHS08
 :GHS07
CAS Number:110-80-5
 :MFCD00008590
Formula:C₂H₅OCH₂CH₂OH
MDL Number:MFCD00002869