Calcium oxide, powder, Laboratory Reagent, J.T.Baker®
Catalog # 1414-R
Supplier: Avantor
CAS Number:
Specifications
- Pk:100 lb
- Seq:7167
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 90 %
- Assay (CaO):≥ 90 %
- Insoluble Matter:≤ 1.0 %
- Loss on Ignition:≤ 2.0 %
- Carbonate (CO₃):Passes Test
- Magnesium and Alkali Salts:Passes Test
Specification Test Results
About this item
:
:56.08 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-064-4
:GHS07
CAS Number:1305-78-8
Formula:CaO
MDL Number:MFCD00010911