Order Entry
Spain
ContactUsLinkComponent
Silver trifluoromethanesulphonate for synthesis, Sigma-Aldrich®
Corrosive
Silver trifluoromethanesulphonate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Silver trifluoromethanesulphonate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    356 °C
  • Molar Mass:
    256.94 g/mol

Specification Test Results

Related Information

Frequently Bought Together