Order Entry
Spain
ContactUsLinkComponent
Trifluoromethanesulphonic acid for synthesis
Trifluoromethanesulphonic acid for synthesis
Supplier:  Merck
Certificates
Trifluoromethanesulphonic acid for synthesis
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble,(caution ! development of heat)
  • Molar Mass:
    150.08 g/mol
  • Boiling Point:
    162 °C (1013 hPa)
  • Vapor Pressure:
    10 hPa (55 °C)
  • Density:
    1.71 g/cm3 (20 °C)
  • pH:
     - 1 (H2O)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together