

Trifluoromethanesulphonic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1493-13-6
:
Formula:CHF₃O₃S
MDL Number:MFCD00007514
:150.08 g/mol
Merck Index:14,9676
Boiling Pt:161…162 °C
Melting Pt:–40 °C
Density:1.708
Storage Temperature:Ambient
EINECS:216-087-5
UN:3265
ADR:8,II
:GHS05
:GHS07
Related Information
Frequently Bought Together