


Silver(I) acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:563-63-3
:
Formula:C₂H₃AgO₂
MDL Number:MFCD00012446
:166.91 g/mol
Merck Index:12,08648
Melting Pt:2000 °C
Density:3.26 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-254-9
ADR:8,II
:M-Clarity™ quality level = MQ200
UN:1759
:GHS05
:GHS07
:GHS09
Specification Test Results
- Solubility:10.2 g/l (20 °C)
- Molar Mass:166.91 g/mol
- Density:3.26 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together