Methyl stearate
Catalog #
Product Details & Documents
Synonyms: Stearic acid methyl ester, Methyl octadecanoate
- CAS Number:112-61-8
- Formula:CH₃(CH₂)₁₆CO₂CH₃
- MDL Number:MFCD00009005
- Molecular Weight:298.51 g/mol
- Merck Index:12,08959
- Boiling Pt:355 °C (1013 hPa)
- Melting Pt:37…39 °C
- Density:1.007 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:203-990-4



