Propionamide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-05-0
:
Formula:C₃H₇NO
MDL Number:MFCD00008039
:73.09 g/mol
Merck Index:12,08009
Boiling Pt:213 °C (1013 hPa)
Melting Pt:79 °C
Density:1.1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:201-172-1
:GHS07
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:75 - 78 °C
- Molar Mass:73.1 g/mol
- Boiling Point:213 °C
- Hill Formula:C3H7NO
- Chemical Formula:CH3CH2CONH2
Specification Test Results
Frequently Bought Together