N,N-Dimethylformamide ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
About this item
- ≥95% for general laboratory use.
Beilstein Number: 605365
RTECS Number: LQ2100000
UNSPSC Code: 12191602
Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
- CAS Number:68-12-2
- Formula:HCON(CH₃)₂
- MDL Number:MFCD00003284
- Molecular Weight:73.09 g/mol
- Merck Index:13,03269
- Boiling Pt:152…153 °C (1.013 hPa)
- Boiling Pt:152-153 °C (1.013 hPa)
- Melting Pt:–61 °C
- Melting Pt:-61 °C
- Density:0.94 g/cm³ (20 °C)
- Flash Pt:57.5 °C (1.013.25 hPa)
- Storage Temperature:Ambient
- EINECS:200-679-5
- UN:2265
- ADR:3,III
- REACH:01-2119475605-32
2 Options Available Below
Product Details & Documents
- ≥95% for general laboratory use.
Beilstein Number: 605365
RTECS Number: LQ2100000
UNSPSC Code: 12191602
Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
- CAS Number:68-12-2
- Formula:HCON(CH₃)₂
- MDL Number:MFCD00003284
- Molecular Weight:73.09 g/mol
- Merck Index:13,03269
- Boiling Pt:152…153 °C (1.013 hPa)
- Boiling Pt:152-153 °C (1.013 hPa)
- Melting Pt:–61 °C
- Melting Pt:-61 °C
- Density:0.94 g/cm³ (20 °C)
- Flash Pt:57.5 °C (1.013.25 hPa)
- Storage Temperature:Ambient
- EINECS:200-679-5
- UN:2265
- ADR:3,III
- REACH:01-2119475605-32
Documents
sds_D158550
SDS
Specifications for Product Family
Specification Test Results
- Autoignition Temperature:833 °F
- Identity (IR):complying
- Assay (GC):Min. 99.0 %
- Water (Karl Fischer):Max. 0.1 %
- Vapor Pressure:2.7 mmHg ( 20 °C)
- Explosion Limit:15.2 %
- Refractive Index:n20/D 1.430 (lit.)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



