Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Acetyl chloride ≥99%

Flammable
Corrosive
Acetyl chloride ≥99%
Certificates
Acetyl chloride ≥99%
Acetyl chloride ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99+% CAS No.: 75-36-5 Molecular Formula: C2H3ClO Formula Weight: 78.50 Flash Point: 4°(39°F) Physical Form: Liquid Density: 1.104 Refractive Index: 1.3890 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Pungent Melting Point: -112° Boiling Point: 50-52° Beilstein Reference: 605303 Merck Reference: 14,85 UN No.: 1717 MDL No.: MFCD00000719

Synonyms:  Acetyl chloride 1M in Dichloromethane
  • CAS Number:75-36-5
  • Formula:C₂H₃ClO
  • MDL Number:MFCD00000719
  • Molecular Weight:78.5 g/mol
  • Merck Index:14,85
  • Boiling Pt:50…52 °C
  • Melting Pt:–112 °C
  • Density:1.104
  • Flash Pt:4 °C (39 °F)
  • Storage Temperature:Ambient
  • EINECS:200-865-6
  • UN:1717
  • ADR:3,II
Product Family Options

Product Information