

Bromoacetyl chloride ≥85.0% (by GC)
: TCI
:22118-09-8
:
:BrCH₂COOCl
:MFCD00000724
:157.39 g/mol
:Refrigerator
:244-790-7
:3265
:8,II
:GHS05
:GHS07
- Appearance:Colorless to Yellow clear liquid
- Purity (GC):min. 85.0 %
Frequently Bought Together