D-(+)-Biotin ≥97.5%, white crystalline powder cell culture reagent
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:58-85-5
:
Formula:C₁₀H₁₆N₂O₃S
MDL Number:MFCD00005541
:244.31 g/mol
Merck Index:13,01231
Boiling Pt:573.6 °C (760 mmHg)
Melting Pt:223…233 °C
Density:1.267 g/cm³ (lit.)
Storage Temperature:Refrigerator
EINECS:200-399-3
Specification Test Results
- Identity Test:Passes
- Purity:≥97.5%
- FTIR:Conforms to Standard
- Solubility (0.02% aq soln):Clear Solution
- Reaction (0.02% aq soln):Very Slightly Acid to Litmus
- Optical Rotation (c=2, 0.1 N NaOH):+89 to +93°
- Organic Volatile Impurities:Passes
Specification Test Results
Frequently Bought Together