L(-)-Tyrosine
Catalog # A3401.1000
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:181.19 g/mol
Merck Index:13,09907
Boiling Pt:351 °C (1013 hPa)
Melting Pt:285 °C
Density:1.41 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-460-4
CAS Number:60-18-4
Formula:C₉H₁₁NO₃
MDL Number:MFCD00002606