Order Entry
Spain
ContactUsLinkComponent
L(+)-Isoleucine for synthesis, Sigma-Aldrich®
L(+)-Isoleucine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
L(+)-Isoleucine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    32.1 g/l (20 °C)
  • Melting Point:
    279 - 280 °C (decomposition)
  • Molar Mass:
    131.17 g/mol
  • Bulk Density:
    280 kg/m3
  • pH:
    6.0 (10 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together