Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',4,4',5,6-Octachlorobiphenyl (PCB No. 195) 100 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,3',4,4',5,6-Octachlorobiphenyl (PCB No. 195) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-195-CS
CAS Number:  
Organic reference standard, 2,2',3,3',4,4',5,6-Octachlorobiphenyl (PCB No. 195) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-195-CS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    52663-78-2
  • Description:
    2,2',3,3',4,4',5,6-Octachlorobiphenyl (PCB No. 195) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₂Cl₈
  • Description:
    2,2',3,3',4,4',5,6-Octachlorobiphenyl (PCB No. 195)

Specifications

About this item