L(+)-Glutamic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:56-86-0
:
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number:MFCD00002634
:147.13 g/mol
Merck Index:13,04482
Boiling Pt:325 °C (1013 hPa)
Melting Pt:160 °C
Density:1.538 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-293-7
Specification Test Results
- Solubility:11.1 g/l (25 °C)
- Melting Point:160 °C
- Molar Mass:147.13 g/mol
- Bulk Density:460 kg/m3
- Density:1.54 g/cm3 (20 °C)
- pH:3.0 - 3.5 (8.6 g/l, H2O, 25 °C)
Specification Test Results
Frequently Bought Together