Order Entry
Spain
ContactUsLinkComponent
 
Organic reference standard, 2,3,4,4',5-Pentachlorobiphenyl (PCB No. 114) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-114-CS
 :  
Organic reference standard, 2,3,4,4',5-Pentachlorobiphenyl (PCB No. 114) 100 µg/ml in isooctane, CIL
  PROOCIL-PCB-114-CS
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    74472-37-0
  • Description:
    2,3,4,4',5-Pentachlorobiphenyl (PCB No. 114) 100 µg/ml in isooctane
  • Environmentally Preferable:
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,3,4,4',5-Pentachlorobiphenyl (PCB No. 114)