Ácido L(+)-glutámico 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: E, Glu, H-Glu-OH, Ácido (S)-(+)-glutámico
- CAS Number:56-86-0
- Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
- MDL Number:MFCD00002634
- Molecular Weight:147.13 g/mol
- Merck Index:13,04482
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:205 °C (desc.)
- Density:1,538 g/cm³ (20 °C)
- Storage Temperature:Ambiente
- EINECS:200-293-7
4 Options Available Below
Product Details & Documents
Synonyms: E, Glu, H-Glu-OH, Ácido (S)-(+)-glutámico
- CAS Number:56-86-0
- Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
- MDL Number:MFCD00002634
- Molecular Weight:147.13 g/mol
- Merck Index:13,04482
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:205 °C (desc.)
- Density:1,538 g/cm³ (20 °C)
- Storage Temperature:Ambiente
- EINECS:200-293-7
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 3 hrs)
- Sulfated ash:≤0.1 %
- Specific optical rotation:+30.5° to +32.5° (20°C, 589 nm) (c=10, 2N HCl) (on dried substance)
- Titration with NaOH:98.5 to 100.5 % (On dried substance)
- Chloride (Cl):≤200 ppm
- Sulfate (SO4):≤300 ppm
- Ammonium (NH4):≤200 ppm
- Iron (Fe):≤30 ppm
- Arsenic oxide (As2O3):≤1 ppm
- Heavy metals (as Pb):≤10 ppm
- Other amino acids:not detectable
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



