Order Entry
Spain
ContactUsLinkComponent
 
Organic reference standard, 2,2',5-Trichlorobiphenyl (PCB No. 18) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-18
 :  
Organic reference standard, 2,2',5-Trichlorobiphenyl (PCB No. 18) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-18
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    37680-65-2
  • Description:
    2,2',5-Trichlorobiphenyl (PCB No. 18) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    2,2',5-Trichlorobiphenyl (PCB No. 18)