Organic reference standard, 2,2',3,5-Tetrachlorobiphenyl (PCB No. 43) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-43
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:70362-46-8
- Description:2,2',3,5-Tetrachlorobiphenyl (PCB No. 43) 35 µg/ml in isooctane
- Environmentally Preferable:
- Formula:C₁₂H₆Cl₄
- Description:2,2',3,5-Tetrachlorobiphenyl (PCB No. 43)