Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, 2,2',3,4,5'-Pentachlorobiphenyl (PCB No. 87) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,4,5'-Pentachlorobiphenyl (PCB No. 87) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-87
CAS Number:  
Organic reference standard, 2,2',3,4,5'-Pentachlorobiphenyl (PCB No. 87) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-87
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    38380-02-8
  • Description:
    2,2',3,4,5'-Pentachlorobiphenyl (PCB No. 87) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,2',3,4,5'-Pentachlorobiphenyl (PCB No. 87)

Specifications

About this item