Organic reference standard, 2,2',3,4,5,5'-Hexachlorobiphenyl (PCB No. 141) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-141
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:52712-04-6
- Description:2,2',3,4,5,5'-Hexachlorobiphenyl (PCB No. 141) 35 µg/ml in isooctane
- Environmentally Preferable:
- Formula:C₁₂H₄Cl₆
- Description:2,2',3,4,5,5'-Hexachlorobiphenyl (PCB No. 141)