Organic reference standard, 2,5-Dichlorobiphenyl (PCB No. 9) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-9
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:34883-39-1
- Description:2,5-Dichlorobiphenyl (PCB No. 9) 35 µg/ml in isooctane
- Formula:C₁₂H₈Cl₂
- Description:2,5-Dichlorobiphenyl (PCB No. 9)